Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94 to 96%, Alfa Aesar™


Overview
Brand: Alfa Aesar

Manufacturer Part Number: 033361.M1

UNSPSC: 12191601

Code: NEW

Additional Details:
Additional Details: CAS Number: 64-17-5 Weight: 0.78900kg Transport: UN number : 1987 Chem class : 2 Pack group : II


Quantity 4 x 1L

Product Code. 15436115

Quantity Price
1 £ 118.72 / 1 L
Estimated Shipment date
from Supplier 13-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Ethanol, Alcohol Reagent, anhydrous
Name Note For analysis ACS
CAS 64-17-5
Physical Form Liquid
Assay Percent Range 94 to 96%
Molecular Formula C2H6O
Formula Weight 46.08
MDL Number MFCD00003568

Specification

Formula Weight 46.08
Quantity 4 x 1L
EINECS Number 200-578-6
Density 0.789g/mL
CAS 64-17-5
Molecular Formula C2H6O
Name Note For analysis ACS
Assay Percent Range 94 to 96%
Physical Form Liquid
MDL Number MFCD00003568
Chemical Name or Material Ethanol, Alcohol Reagent, anhydrous
Documentation

Technical documents