missing translation for 'onlineSavingsMsg'
Learn More

Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Catalog Number 10100332
826.60 GBP valid until 2024-12-31
Use promo code "21953" to get your promotional price.


819.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


819.46 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


826.60 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Metal can
Plastic Coated Glass bottle
Plastic container
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.069
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
CHEBI:16236
ethanol
CCO
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.069
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
CHEBI:16236
ethanol
CCO

C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO

TRUSTED_SUSTAINABILITY
Packaging Metal can
Quantity 25 L
Color Colorless
Formula Weight 46.07g/mol
pH 7
Melting Point -114°C
Boiling Point 78°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Ethanol 96%, technical, duty free
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'