missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™

Meets ACS Specifications

Brand:  Fisher Bioreagents BP2818-100

Code : 32

Additional Details : CAS Number : 64-17-5 Weight : 0.13000kg Transport : UN number : 1170 Chem class : 1 Pack group : II


Quantity 100mL
Packaging Glass Bottle

 View more versions of this product

Product Code. 10517694

  • £18.75 / 100mL


EU Stock 72
Add to basket


Description

Description

  • Meets ACS Specifications
  • For nucleic acid purification and precipitation
  • Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
  • It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding
Specifications

Chemical Identifiers

Ethanol
64-17-5
DNase-, RNase- and Protease-Free
≥99.5%
C2H6O
46.07
Absolute (200 Proof)
Molecular Biology Grade
Liquid
Assay: ≥99.5% (v/v)
C2H5OH
Ethyl Alcohol

Specifications

Ethanol
64-17-5
Molecular Biology Grade
Liquid
Assay: ≥99.5% (v/v)
C2H5OH
2ppm max.
Ethyl Alcohol
DNase free
Conforms to reference
Glass Bottle
0.001% max.
Pass Test
0.0005 mEq /g max.
0.2% max.
12°C (53.6°F)
Clear Colorless
100mL
Absolute (200 Proof)
Pass Test
DNase-, RNase- and Protease-Free
≥99.5%
C2H6O
46.07
15, 3814
For nucleic acid purification and precipitation. Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins. It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding.
Pass Test
0.1% max.
Protease free
Pass Test
Pass Test
0.0002 mEq /g max.
Not Detected
78°C
−114°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS Documentation Special Offers

Special Offers