missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™

Meets ACS Specifications

Brand:  Fisher Bioreagents BP2818-4

Code : 32

Additional Details : CAS Number : 64-17-5 Weight : 3.21000kg Transport : UN number : 1170 Chem class : 3 Pack group : II


Packaging Glass Bottle
Quantity 4L

 View more versions of this product

Product Code. 10041814

  • £197.71 / 4 L

In Stock 45

Please to purchase this item. Need a web account? Register with us today!



Description

Description

  • Meets ACS Specifications
  • For nucleic acid purification and precipitation
  • Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
  • It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding
Specifications

Chemical Identifiers

64-17-5
46.069
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CHEBI:16236
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol

Specifications

Ethanol
64-17-5
DNase-, RNase- and Protease-Free
C2H6O
15, 3814
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
Liquid
Molecular Biology
DNase free
Conforms to reference
Glass Bottle
0.001% max.
Pass Test
0.0005 mEq /g max.
0.2% max.
12°C (53.6°F)
Colorless
4L
Absolute (200 Proof)
Pass Test
Assay: ≥99.5% (v/v)
C2H5OH
2ppm max.
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.07
≥99.5%
For nucleic acid purification and precipitation. Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins. It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding.
Pass Test
0.1% max.
Protease free
Pass Test
Pass Test
0.0002 mEq /g max.
Not Detected
78°C
-114°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS Documents