missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 99%+, Absolute, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10680993
101.68 GBP valid until 2024-12-31
Use promo code "21953" to get your promotional price.


101.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


101.56 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


101.68 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic Bottle
Plastic Container
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO

Ethanol 99%+, Absolute, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Melting Point -114°C
Color Colorless
Boiling Point 78°C
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 59 hPa at 20°C
Viscosity 1.2 mPaS at 20°C
Molecular Weight (g/mol) 46.069
Formula Weight 46.07g/mol
compliance-icons compliance-icons
Product Identifier
  • Ethanol 99+%, absolute
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'