missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol 96% v/v, Certified AR for analysis, meets analytical specification of BP and Ph. Eur., Fisher Chemical™
920.91 GBP valid until 2024-12-31
Use promo code "21953" to get your promotional price.
903.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "21953" to get your promotional price.
903.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Description
C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, -114 deg.C, 3568, Amber glass bottle, Liquid, 702
Specifications
Melting Point | -114°C |
Color | Colorless |
Formula Weight | 46.07g/mol |
Boiling Point | 78°C |
Quantity | 25 L |
Packaging | Metal Can |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Ethanol (96% v/v)
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'