missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 96% v/v, Certified AR for analysis, meets analytical specification of BP and Ph. Eur., Fisher Chemical™

Catalog Number 10332562
316.65 GBP valid until 2024-12-31
Use promo code "21953" to get your promotional price.


235.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


235.62 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.07
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
CHEBI:16236
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
C2H6O
46.07
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
CHEBI:16236

C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, -114 deg.C, 3568, Amber glass bottle, Liquid, 702

TRUSTED_SUSTAINABILITY
Melting Point -114°C
Color Colorless
Boiling Point 78°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Formula Weight 46.07g/mol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'