missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 95% (v/v), Certified AR for Analysis, Fisher Chemical™

Catalog Number p-7073011
196.62 GBP valid until 2024-12-31
Use promo code "21953" to get your promotional price.


158.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


196.62 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
CHEBI:16236
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
64-17-5
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
CHEBI:16236
Packaging Amber glass bottle
Quantity 1 L
Color Colorless
Molecular Formula C2H6O
Formula Weight 46.07g/mol
Melting Point -114°C
Boiling Point 78°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Ethanol (95% v/v)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'