missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethanesulfonic acid, 96%

Brand:  Alfa Aesar™ 014891.18

Code : NEW

Additional Details : CAS Number : 594-45-6 Weight : 0.05000kg Transport : UN number : 2586 Chem class : 1a Pack group : III

Quantity 50g

 View more versions of this product

Product Code. 11317098

  • £88.30 / 50g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Ethanesulfonic acid
110.13
UN2586
Ethylsulfonic acid
594-45-6
MFCD00007529
1743071

Specifications

Ethanesulfonic acid
Liquid
110.13
UN2586
Ethylsulfonic acid
1.343g/mL
−17°C
50g
594-45-6
96%
MFCD00007529
1743071
1.4350
122° to 123°C (0.01mmHg)
Colorless to yellow
>110° (230°F)
Videos
SDS