missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonic acid, 95%, Thermo Scientific Chemicals

Catalog Number 10218433
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
594-45-6
C2H5O3S
109.12
MFCD00007529
CCIVGXIOQKPBKL-UHFFFAOYSA-M
ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
11668
CHEBI:42465
CCS([O-])(=O)=O
594-45-6
C2H5O3S
109.12
MFCD00007529
CCIVGXIOQKPBKL-UHFFFAOYSA-M
ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
11668
CHEBI:42465
CCS([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -17.0°C
Color Yellow to Brown
Density 1.3430g/mL
Boiling Point 136°C to 140°C (3.0 hPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula C2H5SO3H
Refractive Index 1.4325 to 1.4355
Quantity 100 g
Beilstein 04, 5
Specific Gravity 1.343
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
IUPAC Name ethanesulfonate
Formula Weight 110.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethanesulfonic acid, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Ethanesulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'