missing translation for 'onlineSavingsMsg'
Learn More

Ethanediol, Certified AR for Analysis, Fisher Chemical™

Catalog Number 10284430
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Plastic container
This item is not returnable. View return policy
107-21-1
C2H6O2
62.068
2885
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O
This item is not returnable. View return policy
107-21-1
C2H6O2
62.068
2885
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O

C2H6O2, CAS Number-107-21-1, 1,2-ethanediol, ethylene alcohol, ethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, monoethylene glycol, glycol alcohol, glycol, tescol, fridex, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885

TRUSTED_SUSTAINABILITY
Melting Point -13°C
Color Colorless
pH 5.5 to 7.5
Boiling Point 196°C
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Vapor Density 2.14
Formula Weight 62.06g/mol
Vapor Pressure 0.12mmHg at 20°C
Physical Form Viscous Liquid
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'