missing translation for 'onlineSavingsMsg'
Learn More

Ethanediol, Certified AR for Analysis, Fisher Chemical™

C2H6O2, CAS Number-107-21-1, 1,2-ethanediol, ethylene alcohol, ethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, monoethylene glycol, glycol alcohol, glycol, tescol, fridex, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885

Brand:  Fisher Chemical E/0352/17

Code : 22

Additional Details : CAS Number : 107-21-1 Weight : 2.50000kg


Packaging Amber glass bottle
Quantity 2.5L

 View more versions of this product

Product Code. 10040160

  • £177.35 / 2.50 L

In stock, ready to ship: 68

Please to purchase this item. Need a web account? Register with us today!


Description

Description

C2H6O2, CAS Number-107-21-1, 1,2-ethanediol, ethylene alcohol, ethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, monoethylene glycol, glycol alcohol, glycol, tescol, fridex, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885

Specifications

Chemical Identifiers

107-21-1
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
ethane-1,2-diol
C2H6O2
2885
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
CHEBI:30742
C(CO)O

Specifications

Amber glass bottle
0.12mmHg at 20°C
Colorless
2.5L
107-21-1
2885
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
174
62.06g/mol
2.14
196°C
-13°C
5.5 to 7.5
C2H6O2
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
C(CO)O
62.068
CHEBI:30742
Viscous liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethylene glycol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

SDS Documents
Certificates