Learn More
Cytiva ETFE Ferrule
Ferrules used for chromatography tubing. Cytiva Lifescience™ ETFE Ferrule is made from ethylenetetra fluoroethylene and comes with different outside diameters.
Brand:  Cytiva 18-1127-06
Code : GH
Additional Details : Weight : 0.00200kg
 View more versions of this product
Product Code. 11300262
  • £101.00 / Each
Estimated Shipment: 02-05-2024
to see stock.
Add to basket
Description
Description
  • Ferrules
  • Chromatography accessory
  • Used for: protecting the end of tubing
  • Packsize: 10 Connections
  • Ethylenetetra fluoroethylene (ETFE) 
Specifications
Specifications
Tubing
ETFE
10 Pack
1/16 in.
Ferrule