missing translation for 'onlineSavingsMsg'
Learn More

Etazolate hydrochloride, Tocris Bioscience™

PDE4 inhibitor

Brand:  Tocris (Bio-Techne) 043850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13447653

  • £424.83 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Phosphodiesterase inhibitor, selective for PDE4 (IC50 = 2.0 μM).
Specifications

Chemical Identifiers

35838-58-5
325.797
etazolate hydrochloride, etazolate hcl, unii-7yo3254y6b, etazolate hydrochloride usan, dsstox_cid_25767, dsstox_rid_81109, dsstox_gsid_45767, 1-ethyl-4-1-methylethylidene hydrazino 1h-pyrazolo 3,4-b pyridine-5-carboxylic acid ethyl ester hydrochloride, ethyl 1-ethyl-4-2-propan-2-ylidenehydrazinyl pyrazolo 3,4-b pyridine-5-carboxylate hydrochloride
ethyl 1-ethyl-4-(2-propan-2-ylidenehydrazinyl)pyrazolo[3,4-b]pyridine-5-carboxylate;hydrochloride
C14H20ClN5O2
GQJUGJHJUZSJLZ-UHFFFAOYSA-N
37274
CCN1C2=NC=C(C(=C2C=N1)NN=C(C)C)C(=O)OCC.Cl

Specifications

50mg
35838-58-5
etazolate hydrochloride, etazolate hcl, unii-7yo3254y6b, etazolate hydrochloride usan, dsstox_cid_25767, dsstox_rid_81109, dsstox_gsid_45767, 1-ethyl-4-1-methylethylidene hydrazino 1h-pyrazolo 3,4-b pyridine-5-carboxylic acid ethyl ester hydrochloride, ethyl 1-ethyl-4-2-propan-2-ylidenehydrazinyl pyrazolo 3,4-b pyridine-5-carboxylate hydrochloride
CCN1C2=NC=C(C(=C2C=N1)NN=C(C)C)C(=O)OCC.Cl
325.797
325.8
Etazolate hydrochloride
C14H20ClN5O2
GQJUGJHJUZSJLZ-UHFFFAOYSA-N
ethyl 1-ethyl-4-(2-propan-2-ylidenehydrazinyl)pyrazolo[3,4-b]pyridine-5-carboxylate;hydrochloride
37274
>98%
Videos
Documents