Esterase Bs3 From Bacillus Stearothermophilus, Acros Organics


Overview
Brand: Acros Organics

Manufacturer Part Number: 381010250

25MG Esterase BS3 from bacillus stearothermophilus, recombinant in E.Coli, EC 3.1.1.1

UNSPSC: 12352204

Code: 29

Additional Details:
Additional Details: CAS Number: 9016-18-6 Weight: 0.05002kg



Product Code. 10362093

Quantity Price
1 £ 26.8 / 25mg
EU Stock 1
Add to basket


Description and Specification

Specification

Storage Requirements 4°C
Concentration >2.5U/mg
Concentration >2.5U/mg
Color Light Gray
Source Bacillus stearothermophilus
Format Solid
Format Solid
Product Name Esterase BS3
Quantity 25mg
Quantity 25mg
Documentation

Technical documents