missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Esculin sesquihydrate, 97%
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Esculin sesquihydrate is used to inhibits chemically induced carcinogenesis in mouse skin and kidney, used in a microbiology laboratory to aid in the identification of bacterial species, a naturally occurring antioxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Esculin sesquihydrate is used to inhibits chemically induced carcinogenesis in mouse skin and kidney, used in a microbiology laboratory to aid in the identification of bacterial species, a naturally occurring antioxidant.
Solubility
Slightly soluble in water.
Notes
Air Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Esculin sesquihydrate is used to inhibits chemically induced carcinogenesis in mouse skin and kidney, used in a microbiology laboratory to aid in the identification of bacterial species, a naturally occurring antioxidant.
Solubility
Slightly soluble in water.
Notes
Air Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 203°C to 205°C |
Assay Percent Range | 97% |
Quantity | 5 g |
Beilstein | 95387 |
Merck Index | 14,3698 |
Solubility Information | Slightly soluble in water. |
Optical Rotation | −41° (c=5 in pyridine) |
Formula Weight | 367.31 (340.28 Anhydrous) |
Percent Purity | 97% |
Chemical Name or Material | Esculin sesquihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'