missing translation for 'onlineSavingsMsg'
Learn More

Esculin sesquihydrate, Thermo Scientific Chemicals

Catalog Number 15435389
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
66778-17-4
C30H38O21
734.613
MFCD00149492
LGMHQDJZWHRZPO-HPPUHPNBSA-N
esculin sesquihydrate
131675424
7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one;trihydrate
C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.O.O.O
This item is not returnable. View return policy
66778-17-4
C30H38O21
734.613
MFCD00149492
LGMHQDJZWHRZPO-HPPUHPNBSA-N
esculin sesquihydrate
131675424
7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one;trihydrate
C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O.O.O.O

A naturally occuring antioxidant

A naturally occuring antioxidant

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 202°C to 210°C
Color White
Quantity 100 g
Merck Index 14,3698
Formula Weight 367.31 (340.29 Anhydrous)
Physical Form Powder
Chemical Name or Material Esculin sesquihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'