missing translation for 'onlineSavingsMsg'
Learn More

erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide, 95%, Thermo Scientific Chemicals

Catalog Number p-7034415
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
388071-27-0
C15H19F2NO3
299.32
MFCD08061630
NKGKCDXMOMAORK-UHFFFAOYNA-N
erythro-n-boc-l-3,5-difluorophenylalanine epoxide, tert-butyl s-2-3,5-difluorophenyl-1-s-oxiran-2-yl ethyl carbamate, tert-butyl n-1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethyl carbamate, erythro-n-boc-3,5-difluoro-l-phenylalanine epoxide, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiranyl ethylcarbamate, tert-butyl s-2-3,5-difluorophenyl-1-s-oxiran-2-yl ethylcarbamate, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethylcarbamate, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethyl carbamate
9922319
tert-butyl N-[(1S)-2-(3,5-difluorophenyl)-1-[(2S)-oxiran-2-yl]ethyl]carbamate
CC(C)(C)OC(=O)NC(CC1=CC(F)=CC(F)=C1)C1CO1
This item is not returnable. View return policy
388071-27-0
C15H19F2NO3
299.32
MFCD08061630
NKGKCDXMOMAORK-UHFFFAOYNA-N
erythro-n-boc-l-3,5-difluorophenylalanine epoxide, tert-butyl s-2-3,5-difluorophenyl-1-s-oxiran-2-yl ethyl carbamate, tert-butyl n-1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethyl carbamate, erythro-n-boc-3,5-difluoro-l-phenylalanine epoxide, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiranyl ethylcarbamate, tert-butyl s-2-3,5-difluorophenyl-1-s-oxiran-2-yl ethylcarbamate, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethylcarbamate, tert-butyl 1s-2-3,5-difluorophenyl-1-2s-oxiran-2-yl ethyl carbamate
9922319
tert-butyl N-[(1S)-2-(3,5-difluorophenyl)-1-[(2S)-oxiran-2-yl]ethyl]carbamate
CC(C)(C)OC(=O)NC(CC1=CC(F)=CC(F)=C1)C1CO1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 299.31
Percent Purity 95%
Chemical Name or Material erythro-N-Boc-3,5-difluoro-L-phenylalanine epoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'