missing translation for 'onlineSavingsMsg'
Learn More

(±)-erythro-Aleuritic acid, 95%, ACROS Organics™

Brand:  Acros Organics 133730250

Code : 29

Additional Details : CAS Number : 533-87-9 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10255040

  • £72.50 / 25g


EU Stock 7
Add to basket

Specifications

Chemical Identifiers

(±)-erythro-Aleuritic acid
533-87-9
100.0
95%
HO(CH2)5CH(OH)CH(OH)(CH2)7COOH
MFCD00135596
95%
94.0
≥94%
C16H32O5
304.41
9, 10, 16-Trihydroxypalmitic acid

Specifications

(±)-erythro-Aleuritic acid
533-87-9
100.0
Glass bottle
≥94%
C16H32O5
304.41
03, III, 901
9, 10, 16-Trihydroxypalmitic acid
25g
95%
94.0
Authentic
Crystalline Powder
95%
HO(CH2)5CH(OH)CH(OH)(CH2)7COOH
MFCD00135596
15, 225
White to beige
Videos
SDS