missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Erythrina Christagalli Agglutinin (ECA)

Naturally occuring lectin with affinity for D-galactose and D-galactosides

Brand:  Alfa Aesar™ J60146.MA

Code : NEW

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15442969

  • £221.12 / 10mg


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Specifications

White
Powder
10mg
Naturally occuring lectin with affinity for D-galactose and D-galactosides
Agglutinin
−30° to −10°C
Videos
SDS