missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Erlenmeyer Shape With Assembly Set Filtering Flask

Erlenmeyer Shape. DURAN™ Erlenmeyer Shape With Assembly Set Filtering Flask includes KECK assembly kit.

Brand:  Duran™ 212046351

Code : A0

Additional Details : Weight : 1.35000kg

 View more versions of this product

Product Code. 10768321

  • £111.00 / Each


Estimated Shipment
08-02-2018
Add to basket

Description

Description

  • DURAN™ Filtering flasks
  • Erlenmeyer shape
  • With KECK assembly set
Specifications

Specifications

Yes
166mm
Borosilicate Glass
DIN 12 476, ISO 6556, USP Standard
2000mL
255mm
Videos