missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Erlenmeyer Shape Side Arm Socket Filtering Flask

Brand:  Duran™ 211836309

Code : A0

Additional Details : Weight : 1.35000kg

 View more versions of this product

Product Code. 10747551

  • £143.00 / Each


Estimated Shipment
25-06-2018
Add to basket

Specifications

Specifications

Yes
165mm
Borosilicate Glass
DIN 12 476, ISO 6556, USP Standard
2000mL
255mm
Videos