missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Erlenmeyer Shape Side Arm Socket Filtering Flask

Brand:  Duran™ 211835407

Code : A0

Additional Details : Weight : 7.70000kg

 View more versions of this product

Product Code. 11850560

  • £570.00 / Pack of 10


Estimated Shipment
06-02-2018
Add to basket

Specifications

Specifications

Yes
135mm
Borosilicate Glass
DIN 12 476, ISO 6556, USP Standard
1000mL
230mm
Videos