Learn More
Duran™ Erlenmeyer Shape Glass Hose Connection Filtering Flask
Manufactured of Borosilicate Glass. DURAN™ Erlenmeyer Shape Glass Hose Connection Filtering Flask include hose connector.
Brand:  Duran™ 212014401
Code : A0
Additional Details : Weight : 0.47000kg
 View more versions of this product
Product Code. 10412951
  • £37.45 / Each
In Stock 2
Add to basket
Description
Description
  • DURAN® Filtering flask
  • Erlenmeyer shape
  • With glass hose connection
Specifications
Specifications
Yes
105mm
Borosilicate Glass
ISO 6556, USP Standard
500mL
185mm
Videos