Erlenmeyer Flask Clamps for Sartorius™ Certomat™ Shakers
Available in stainless steel or plastic for a variety of flask sizes
Overview
Brand: Sartorius Stedim Biotech
Manufacturer Part Number: BBI-8854711
PINCE PLASTIQUE FOR ERLEN 250ML
UNSPSC: 41121710
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11790406
Quantity Price
1 £ 20.45 / Each
Estimated Shipment
03-04-2017
Add to basket
Description and Specification
Specification
Material Reinforced Plastic
Type Clamp
Capacity (Metric) 250mL
Holds 250mL Erlenmeyer Flask
For Use With Certomat™ MOII RM SII IS, BS-1, BS-T, CT Plus