Erlenmeyer Flask Clamps for Sartorius™ Certomat™ Shakers

Available in stainless steel or plastic for a variety of flask sizes

Overview
Brand: Sartorius Stedim Biotech

Manufacturer Part Number: BBI-8854610

SS CLAMP FOR ERLEN 2000ML

UNSPSC: 41121710

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11740406

Quantity Price
1 £ 39.63 / Each
Estimated Shipment
03-04-2017
Add to basket


Description and Specification

Specification

Material Stainless Steel
Type Clamp
Capacity (Metric) 2L
Holds 2000mL Erlenmeyer Flask
For Use With Certomat™ MOII RM SII IS, BS-1, BS-T, CT Plus