Erlenmeyer Flask Clamps for Sartorius™ Certomat™ Shakers

Available in stainless steel or plastic for a variety of flask sizes

Overview
Brand: Sartorius Stedim Biotech

Manufacturer Part Number: BBI-8854572

Clamps for 500ml flasks

UNSPSC: 41100000

Additional Details:
Additional Details: Weight: 5.00000kg



Product Code. 12767126

Quantity Price
1 £ 31.96 / Each
Estimated Shipment
06-03-2017
Add to basket


Description and Specification

Specification

Material Stainless Steel
Type Clamp
Capacity (Metric) 500mL
Holds 500mL Erlenmeyer Flask
For Use With Certomat™ MOII RM SII IS, BS-1, BS-T, CT Plus