ERK 1/2 Mouse, eFluor 570, Clone: 5AD13MA, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 41-9108-80

Code: NEW

Additional Details:
Additional Details: Weight: 0.09500kg



Disclaimers: For Research Use Only.

Product Code. 15598946

Quantity Price
1 £ 67.82 / 25µg
Estimated Shipment date
from Supplier 05-05-2017
Add to basket


Description and Specification

Specification

Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Primary or Secondary Primary
Conjugate eFluor 570
Host Species Mouse
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Regulatory Status RUO
Antigen ERK 1/2
Quantity 25μg
Species Reactivity Human, Mouse, Rat
Clone 5AD13MA
Gene Alias MAPK, ERK, p42/p44
Format Conjugated
Concentration 0.2mg/mL
Isotype IgG1
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.

The 5AD13MA monoclonal antibody recognizes human, mouse, and rat extracellular signal-regulated kinases 1 and 2 (also known as ERK1/2, p44/42, or MAPK3/1). ERK1/2 belong to a family of conserved serine/threonine protein kinases known as mitogen-activated protein kinases (MAPKs) that are involved in many cellular programs such as proliferation, differentiation, motility, and survival. The ERK1/2 signaling pathway can be activated in response to a diverse range of extracellular stimuli including mitogens, growth factors, and cytokines. The primary activators of ERK1/2 are MEK1 and MEK2 which activate p44 and p42 through phosphorylation of activation loop residues Thr202/Tyr204 and Thr185/Tyr187 in ERK1 and ERK2, respectively. Several downstream targets of ERK1/2 have been identified, including p90RSK and the transcription factor Elk-1. ERK1/2 are negatively regulated by MAPK phosphatases as well as by chemical inhibitors of MEK including U0126 and PD98059. The ERK pathway is disrupted in many types of cancer and thus is an important target for diagnosis and treatment.