missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) iodide, ultra dry, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7050136
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13813-42-8
MFCD00016075
erbium iii iodide, erbium iii ; iodide, acmc-1bohg, erbium iodide,
This item is not returnable. View return policy
13813-42-8
MFCD00016075
erbium iii iodide, erbium iii ; iodide, acmc-1bohg, erbium iodide,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1280°C
Melting Point 1020°C
Physical Form Powder or Pieces
Quantity 1 g
Formula Weight 547.97
Percent Purity 99.9%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Erbium(III) iodide, Ultra dry

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'