missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Erbium(III) iodide, anhydrous, 90% min, (REO)

Brand:  Alfa Aesar™ 033540.04

Code : NEW

Additional Details : CAS Number : 13813-42-8 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11377748

  • £37.95 / 2g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Erbium(III) iodide
Anhydrous
MFCD00016075
13813-42-8
547.97

Specifications

Erbium(III) iodide
13813-42-8
≤90%
547.97
Hygroscopic
1020°C
Anhydrous
Crystalline
(REO)
MFCD00016075
1280°C
2g
Videos
SDS