missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) acetate tetrahydrate, 99% (REO), Thermo Scientific Chemicals

Catalog Number p-7059996
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
15280-57-6
C6H9ErO6·4H2O
MFCD00013034
This item is not returnable. View return policy
15280-57-6
C6H9ErO6·4H2O
MFCD00013034

Erbium(III) acetate tetrahydrate is used for laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components, photo-optical material and optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Pink
Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula Er(OOCCH3)3·4H2O
Solubility Information Soluble in water.
Formula Weight 416.45 (344.39 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.114 g/mL
Assay (REO)
Chemical Name or Material Erbium (III) acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'