missing translation for 'onlineSavingsMsg'
Learn More

Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7048990
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-52-0
Er
167.26
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
23980
CHEBI:33379
erbium
[Er]
This item is not returnable. View return policy
7440-52-0
Er
167.26
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
23980
CHEBI:33379
erbium
[Er]

Erbium foil used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films. It is used as an alloying element with titanium and is also used to produce infra-red absorbing glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
UN Number UN3178
Quantity 25 x 25 mm
Solubility Information Insoluble in water.
Percent Purity 99.9%
Weight ≈0.14g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Erbium foil, 0.025mm (0.001 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'