missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ ERBB3 Recombinant Human Protein (without Catalytic Activity Domain), His Tag

Expressed from DNA sequence encoding extracellular domain (Met 1-Thr 643) of human ERBB3 precursor fused to polyhistidine tag

Brand:  Sino Biological™ 10201H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13475806

  • £920.25 / 250µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

  • Activity is measured by its binding ability in functional ELISA
  • Immobilized human ERBB3-Fc at 1μg/well can bind biotinylated recombinant human NRG1 w/linear range of 0.032-0.16μg/mL
  • ED50 of human NRG1 is 0.43 μg/ml
Specifications

Specifications

Studying protein-protein interactions and protein structure analysis
2065
≥85% by SDS-PAGE
Human
Truncated, Cell Surface Protein, Recombinant
V-erb-b2 erythroblastic leukemia viral oncogene homolog 3
Assay Standard
Mammalian
His Tag
Lyophilized
70.2kd
5 x 50μg
−20° to −70°C
<1 EU/μg
ERBB3
Growth Factors and Receptors
ERBB3 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.