missing translation for 'onlineSavingsMsg'
Learn More

ERB 041, Tocris Bioscience™

Potent ERβ agonist

Brand:  Tocris (Bio-Techne) 427610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13434474

  • £204.33 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent ERβ agonist; displays >200-fold selectivity for ERβ over ERα (IC50 values are 5 and 1216 nM for human ERβ and ERα; 3.1 and 620 nM for rat ERβ and ERα; and 3.7 and 750 nM for mouse ERβ and ERα respectively).
Specifications

Chemical Identifiers

524684-52-4
271.247
prinaberel, unii-a9c8mnf7ca, a9c8mnf7ca, 2-3-fluoro-4-hydroxyphenyl-7-vinyl-1,3-benzoxazol-5-ol, prinaberel usan:inn, 4-2z-7-ethenyl-5-hydroxy-3h-1,3-benzoxazol-2-ylidene-2-fluorocyclohexa-2,5-dien-1-one, prinaberel usan/inn, bidd:er0139, erb hplc, 7-ethenyl-2-3-fluoro-4-hydroxyphenyl benzoxazol-5-ol
(4Z)-4-(7-ethenyl-5-hydroxy-3H-1,3-benzoxazol-2-ylidene)-2-fluorocyclohexa-2,5-dien-1-one
C15H10FNO3
FCXYSEXZEGPLGG-DHDCSXOGSA-N
5326893
C=CC1=CC(=CC2=C1OC(=C3C=CC(=O)C(=C3)F)N2)O

Specifications

10mg
524684-52-4
prinaberel, unii-a9c8mnf7ca, a9c8mnf7ca, 2-3-fluoro-4-hydroxyphenyl-7-vinyl-1,3-benzoxazol-5-ol, prinaberel usan:inn, 4-2z-7-ethenyl-5-hydroxy-3h-1,3-benzoxazol-2-ylidene-2-fluorocyclohexa-2,5-dien-1-one, prinaberel usan/inn, bidd:er0139, erb hplc, 7-ethenyl-2-3-fluoro-4-hydroxyphenyl benzoxazol-5-ol
C=CC1=CC(=CC2=C1OC(=C3C=CC(=O)C(=C3)F)N2)O
271.247
271.24
ERB 041
C15H10FNO3
FCXYSEXZEGPLGG-DHDCSXOGSA-N
(4Z)-4-(7-ethenyl-5-hydroxy-3H-1,3-benzoxazol-2-ylidene)-2-fluorocyclohexa-2,5-dien-1-one
5326893
>99%
Videos
Documents