missing translation for 'onlineSavingsMsg'
Learn More

ER 27319 maleate, Tocris Bioscience™

Selective Syk kinase inhibitor

Brand:  Tocris (Bio-Techne) 247110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441284

  • £137.70 / 10mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of Syk kinase. Inhibits tyrosine phosphorylation of Syk initiated by the engagement of FcεRI in rat and human mast cells. Results in the abrogation of degranulation, TNF-α production (IC50 = 10 μM) and other related signaling events.
Specifications

Chemical Identifiers

1204480-26-1
396.443
er 27319 maleate, 10-3-aminopropyl-3,4-dimethyl-9 10h-acridinone maleate
10-(3-aminopropyl)-3,4-dimethylacridin-9-one;(Z)-but-2-enedioic acid
C22H24N2O5
WVUQPGFRFBVJKH-BTJKTKAUSA-N
56972172
CC1=C(C2=C(C=C1)C(=O)C3=CC=CC=C3N2CCCN)C.C(=CC(=O)O)C(=O)O

Specifications

ER 27319 maleate
1204480-26-1
er 27319 maleate, 10-3-aminopropyl-3,4-dimethyl-9 10h-acridinone maleate
CC1=C(C2=C(C=C1)C(=O)C3=CC=CC=C3N2CCCN)C.C(=CC(=O)O)C(=O)O
396.443
396.17
10mg
C22H24N2O5
WVUQPGFRFBVJKH-BTJKTKAUSA-N
10-(3-aminopropyl)-3,4-dimethylacridin-9-one;(Z)-but-2-enedioic acid
56972172
>99%
Videos