missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epTIPS™ Pipette Tip Reloads

Use this refill system for worry-free work in the molecular biology lab. Eppendorf™ epT.I.P.S.™ Pipette Tip Reloads allow contamination-free handling of the refill trays. Plastic lids protect the trays from direct contact during transfer to the working box.

Brand:  Eppendorf™ 0030073762

Code : 96

Additional Details : Weight : 0.59130kg

 View more versions of this product

Product Code. 10330272

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Use this refill system for worry-free work in the molecular biology lab. Eppendorf™ epT.I.P.S.™ Pipette Tip Reloads allow contamination-free handling of the refill trays. Plastic lids protect the trays from direct contact during transfer to the working box.

This platinum-cured silicon pump tubing is the best choice for applications involving pressure, suction lift, viscous fluids or long tubing life.
  • Complies with USP Class VI, EP, FDA, 3A, REACH,  RoHS,  ADCF
  • Low absorption; excellent biocompatibility
  • Very low extractable; weather, ozone, corona, radiation resistant
  • Formulation: Tygon™ 3350 silicone, platinum cured
  • Ultra-smooth; minimizes bacterial growth
  • Good for mild to medium concentration bases, salt and alcohols
  • Transparent, odorless, tasteless and nontoxic
Specifications

Specifications

Eppendorf™ Pipettor
Autoclavable
PCR clean
Rack
0.1 to 20 μL
4cm
Eppendorf™
Videos
SDS
Documents
Certificates
Special Offers

Special Offers