Learn More
Eppendorf™ epTIPS™ Pipette Tip Reloads
Use this refill system for worry-free work in the molecular biology lab. Eppendorf™ epT.I.P.S.™ Pipette Tip Reloads allow contamination-free handling of the refill trays. Plastic lids protect the trays from direct contact during transfer to the working box.
Brand: Eppendorf™ 0030073363
Code : 96
Additional Details : Weight : 0.54900kg
Description
Use this refill system for worry-free work in the molecular biology lab. Eppendorf™ epT.I.P.S.™ Pipette Tip Reloads allow contamination-free handling of the refill trays. Plastic lids protect the trays from direct contact during transfer to the working box.
- Contamination free transfer of refill trays to the refill box (epT.I.P.S.™ box) in plastic covers
- Racks can be autoclaved
- Also available in PCR* clean quality, free of human DNA, DNase, RNase and PCR* inhibitors
- 0.1μL to 10μL and 2μL to 200μL tips come in stacks of five that can be autoclaved as a stack
Specifications
0.1 to 10 μL | |
Autoclavable | |
n.a. | |
Universal Fit | |
epT.I.P.S. |
Non-Filtered | |
3.4cm | |
Rack | |
Eppendorf™ Pipettor |