missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ epTIPS™ Low-retention Pipettor Tips
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tips are ideal for cell culture (media) applications and applications with samples that contain detergents.
£35.41
Specifications
Filtered | Non-Filtered |
---|---|
Sterility | Sterile |
For Use With (Application) | They satisfy the most stringent demands in medicine, the pharmaceutical and food industries, PCR*, molecular biology and cytotechnology |
Tip Style | Low Retention |
For Use With (Equipment) | Eppendorf Pipetter |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10035341
|
Eppendorf™
0030010035 |
2 to 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10536252
|
Eppendorf™
0030010051 |
50 to 1000 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tips are ideal for cell culture (media) applications and applications with samples that contain detergents.
- Ultrahomogenous surface for maximum reproducibility
- Ultrahydrophobic surface for minimal sample loss
- Significantly reduced foam formation during pipetting
- Extreme resistance to chemicals
- Available in PCR clean and Eppendorf Quality
- Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free)
- No coating
Specifications
Non-Filtered | |
They satisfy the most stringent demands in medicine, the pharmaceutical and food industries, PCR*, molecular biology and cytotechnology | |
Eppendorf Pipetter |
Sterile | |
Low Retention | |
epT.I.P.S. |