missing translation for 'onlineSavingsMsg'
Learn More

epsilon-Caprolactone, 99%, Thermo Scientific Chemicals

Catalog Number p-7080670
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
502-44-3
C6H10O2
114.144
MFCD00003267
PAPBSGBWRJIAAV-UHFFFAOYSA-N
6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane
10401
CHEBI:17915
oxepan-2-one
C1CCC(=O)OCC1
502-44-3
C6H10O2
114.144
MFCD00003267
PAPBSGBWRJIAAV-UHFFFAOYSA-N
6-hexanolactone, epsilon-caprolactone, 2-oxepanone, caprolactone, hexan-6-olide, 6-hexanolide, hexano-6-lactone, 1,6-hexanolide, e-caprolactone, 1-oxa-2-oxocycloheptane
10401
CHEBI:17915
oxepan-2-one
C1CCC(=O)OCC1

ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ε-Caprolactone is widely used as a monomer in the manufacturing of highly specialized polymers. It is mainly utilized as a precursor to caprolactam. It finds an application in the synthesis of polyglecaprone, which is used as a suture material in surgery.

Solubility
Miscible with water and most organic solvents.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Combustible liquid.
TRUSTED_SUSTAINABILITY
Melting Point -2°C
Density 1.078
Boiling Point 235°C to 236°C
Flash Point 109°C (228°F)
Assay Percent Range 99%
Refractive Index 1.463
Quantity 100 g
Beilstein 106919
Solubility Information Miscible with water and most organic solvents.
Formula Weight 114.14
Percent Purity 99%
Chemical Name or Material epsilon-Caprolactone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'