Learn More
EPS8L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559447
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SVSCPLLSRD AVDFLRGHLV PKEMSLWESL GESWMRPRSE WPREPQVPLY VPKFHSGWEP PVDVLQEAPW EVEGLASAPI EEVSPVSR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
EPS8L2 Polyclonal antibody specifically detects EPS8L2 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
EPS8L2 | |
Polyclonal | |
Unconjugated | |
EPS8L2 | |
AI042819; AW545405; Epidermal growth factor receptor kinase substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-related protein 2; EPS8 like 2; EPS8 related protein 2; Eps8l2; EPS8-like 2; EPS8-like protein 2; EPS8R2; EPS8-related protein 2; PP13181 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
361674, 64787 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6S3 | |
EPS8L2 | |
Recombinant protein corresponding to Human EPS8L2 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Research Use Only