missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Vis Cuvettes

Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm. The Vis cuvettes are the perfect tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD 600 methods), kinetic, and fluorescence measurements.

Brand:  Eppendorf™ 0030079353

Code : NEW

Additional Details : Weight : 2.40000kg

 View more versions of this product

Product Code. 15234566

  • £113.30 / Pack of 1000


Estimated Shipment
25-06-2018
Add to basket

Description

Description

  • Two different cuvette sizes (semi-micro and macro) for a wide variety of volumes and applications
  • Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm
Specifications

Specifications

Plastic
Semi-micro Vis Cuvet
Videos