Learn More
Eppendorf™ Vis Cuvettes
Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm. The Vis cuvettes are the perfect tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD 600 methods), kinetic, and fluorescence measurements.
Brand: Eppendorf™ 0030079345
Code : NEW
Additional Details : Weight : 2.40000kg
Description
- Two different cuvette sizes (semi-micro and macro) for a wide variety of volumes and applications
- Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm
Specifications
Macro Vis Cuvet | |
Plastic |