missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Varispenser™ plus/Varispenser Bottle-Top Reagent Dispensers

Bottle-top reagent dispenser for taking aliquots of liquid from large supply bottles

Brand:  Eppendorf™ 4961000012

Code : 97

Additional Details : Weight : 0.41000kg

 View more versions of this product

Product Code. 10120051

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • PFA-sealing of the slide piston prevents jamming Fine adjustment depending on viscosity of liquid No crystallization of liquid
  • No dripping when valve closed Telescopic tube for use with high bottles Rapid volume setting using precise graduation scale Patented safety valve for deairing without loss of reagent Cannel-sealing cap prevents media contact Autoclavable
  • Extremely chemical-resistant
  • Varispenser plus features safety valve and fine adjustment

Specifications

Specifications

0.5 to 2.5mL
±0.6%
EN ISO 8655
Bottletop Reagent Dispenser
2 Years
Yes
≤0.1%
0.05mL
Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment
Videos
Documentation