missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Varispenser™ plus/Varispenser Bottle-Top Reagent Dispensers

Bottle-top reagent dispenser for taking aliquots of liquid from large supply bottles

Brand:  Eppendorf™ 4960000051

Code : 97

Additional Details : Weight : 0.56000kg

 View more versions of this product

Product Code. 10597951

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • PFA-sealing of the slide piston prevents jamming Fine adjustment depending on viscosity of liquid No crystallization of liquid
  • No dripping when valve closed Telescopic tube for use with high bottles Rapid volume setting using precise graduation scale Patented safety valve for deairing without loss of reagent Cannel-sealing cap prevents media contact Autoclavable
  • Extremely chemical-resistant
  • Varispenser plus features safety valve and fine adjustment

Specifications

Specifications

10 to 50mL
±0.5%
EN ISO 8655
Bottletop Reagent Dispenser
2 Years
Yes
≤0.1%
1mL
Telescopic tube, 3 adapters for 32, 38, 40mm O.D. bottle thread
Videos
Documentation