missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Varispenser™ plus/Varispenser Bottle-Top Reagent Dispensers

Bottle-top reagent dispenser for taking aliquots of liquid from large supply bottles

Brand:  Eppendorf™ 4961000039

UNSPSC : 41121516

Code : 97

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10458502

  • £246.00 / Each


Estimated Shipment
28-07-2017
Add to basket

Specifications

Description

  • PFA-sealing of the slide piston prevents jamming Fine adjustment depending on viscosity of liquid No crystallization of liquid
  • No dripping when valve closed Telescopic tube for use with high bottles Rapid volume setting using precise graduation scale Patented safety valve for deairing without loss of reagent Cannel-sealing cap prevents media contact Autoclavable
  • Extremely chemical-resistant
  • Varispenser plus features safety valve and fine adjustment

Description & Specifications

Specifications

2 to 10mL
±0.5%
EN ISO 8655
Bottletop Reagent Dispenser
2 Years
Yes
≤0.1%
0.2mL
Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment
Documentation Alternative Products