Eppendorf™ Varispenser™ plus/Varispenser Bottle-Top Reagent Dispensers

Bottle-top reagent dispenser for taking aliquots of liquid from large supply bottles

Overview
Brand: Eppendorf™

Manufacturer Part Number: 4961000020

UNSPSC: 41121516

Code: 97

Additional Details:
Additional Details: Weight: 0.38300kg



Product Code. 10518341

Quantity Price
1 £ 246.0 / Each
Estimated Shipment
30-05-2017
Add to basket


Description and Specification

Specification

Product Type Bottletop Reagent Dispenser
Autoclavable Yes
Dispensing Interval 0.1mL
Precision ≤0.1%
Accuracy ±0.5%
Warranty 2 Years
Includes Telescopic tube, 3 adapters for 28, 40, 45mm O.D. bottle thread, recirculation valve, valve toggle and media specific fine adjustment
Volume Range 1 to 5mL
Certifications/Compliance EN ISO 8655

  • PFA-sealing of the slide piston prevents jamming Fine adjustment depending on viscosity of liquid No crystallization of liquid
  • No dripping when valve closed Telescopic tube for use with high bottles Rapid volume setting using precise graduation scale Patented safety valve for deairing without loss of reagent Cannel-sealing cap prevents media contact Autoclavable
  • Extremely chemical-resistant
  • Varispenser plus features safety valve and fine adjustment

Documentation