Learn More
Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc
Fluorescence intensity up to ten times higher than with frosted wells
Brand:  Eppendorf™ 0030132734
Code : A0
Additional Details : Weight : 0.72000kg
 View more versions of this product
Product Code. 11990527
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
The limiting factor in low volume real-time PCR is often the remaining intensity of fluorescence. The white wells of the Eppendorf twin.tec real-time PCR plates reflect fluorescence much better than clear or frosted wells. Thus, lower levels of fluorescence are still detectable with the same instrument–just by using the right consumables. Additionally, white wells significantly reduce interfering background fluorescence and lead to increased homogeneity of replicates and reproducible results.
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121°C, 20 min.)
  • Rigid polycarbonate frame ensures easy and reliable handling, manually or automated
  • Polypropylene wells guarantee excellent and fast temperature transfer to the sample
Specifications
Specifications
Yes
White
Polypropylene
Skirted
White
Yes
RT-PCR applications
Yes
Sterile
Videos