Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc

Fluorescence intensity up to ten times higher than with frosted wells

Overview
Brand: Eppendorf™

Manufacturer Part Number: 0030132726

x20 twintec real-time PCR 96 low profile, blackprof., schwarz: VE=20 Stück

UNSPSC: 41113036

Code: 96

Additional Details:
Additional Details: Weight: 0.59200kg



Product Code. 10217467

Quantity Price
1 £ 95.0 / Pack of 20
Estimated Shipment
04-04-2017
Add to basket


Description and Specification

Specification

Well Color White
Material Polypropylene
Autoclavable Yes
Rimmed Yes
Sterile Yes
Barcode Yes
Product Type RT-PCR Plate
Color Black
Material Polypropylene
Applications RT-PCR applications
For Use With (Application) RT-PCR applications
Skirt Style Non-skirt
Sterile Yes