Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc

Fluorescence intensity up to ten times higher than with frosted wells

Overview
Brand: Eppendorf™

Manufacturer Part Number: 0030132750

x20 twintec real-time PCR 384, Black

UNSPSC: 41113036

Code: A0

Additional Details:
Additional Details: Weight: 0.81000kg



Product Code. 11910537

Quantity Price
1 £ 301.0 / Pack of 25
Estimated Shipment
28-02-2017
Add to basket


Description and Specification

Specification

Well Color White
Material Polypropylene
Autoclavable Yes
Rimmed Yes
Sterile Yes
Barcode Yes
Product Type RT-PCR Plate
Color Black
Material Polypropylene
Applications RT-PCR applications
For Use With (Application) RT-PCR applications
Skirt Style Skirted
Sterile Yes