missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ ThermoMixer™ C Accessory, Smartblock™ Thermoblock

Maintain accurate uniform temperatures with these thermoblocks. Eppendorf™ Smartblock™ Thermoblocks are compatible with Eppendorf ThermoMixer™ C, and support a broad range of applications.

Brand:  Eppendorf™ 5361000031

Code : X3

Additional Details : Weight : 0.58500kg

 View more versions of this product

Product Code. 13588460

  • £443.71 / Each


Estimated Shipment
20-02-2018
Add to basket

Description

Description

  • Up to 24 tubes (2.0mL)
  • Exchangeable in just seconds
  • Automatic block recognition for safe handling

 

Specifications

Specifications

Eppendorf ThermoMixer C
Thermoblock
0.5mL x 24 Reaction vessels
0.5mL x 24 Reaction vessels
Videos